2',3'-O-Isopropylideneadenosine ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
CAS Number: 362-75-4
MDL Number: MFCD00005756
Molecular Formula: C13H17N5O4
Molecular Weight: 307.31
Purity/Analysis Method:
Product Categories: Bioscience, Nucleosides and Their Analogs
Other Product Names / Synonyms: 9-(2,3-O-Isopropylidene-beta-D-ribofuranosyl)adenine
Formula:
C₁₃H₁₇N₅O₄ Melting Pt: 222 °C |
MDL Number:
MFCD00005756 CAS Number: 362-75-4 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...