Iodoethane ≥99.0% stabilized
Supplier: TCI America
|
Danger
|
(stabilized with Copper chip)
CAS Number: 75-03-6
MDL Number: MFCD00001091
Molecular Formula: C2H5I
Molecular Weight: 155.97
Purity/Analysis Method:
Other Product Names / Synonyms: Ethyl Iodide
Formula:
C₂H₅I Boiling Pt: 72 °C Melting Pt: –108 °C Density: 1.94 (20 °C) Flash Pt: 53 °C Storage Temperature: Ambient |
MDL Number:
MFCD00001091 CAS Number: 75-03-6 EINECS: 200-833-1 UN: 2810 ADR: 6.1,III Merck Index: 13,03846 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...