Iodoacetamide ≥98.0% (by total nitrogen basis)

Supplier: TCI America
Warning

I0044-25G I0044-500G I0044-100G
TCI0044-025GEA 79.91 USD
TCI0044-025G TCI0044-500G TCI0044-100G
Iodoacetamide ≥98.0% (by total nitrogen basis)
Iodoacetamide

CAS Number: 144-48-9 MDL Number: MFCD00008028 Molecular Formula: C2H4INO Molecular Weight: 184.96 Purity/Analysis Method: 98.0% (N) Form: Crystal Melting point (°C): 95

Formula: ICH₂CONH₂
Boiling Pt: 266 °C (1013 hPa)
Melting Pt: 95 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00008028
CAS Number: 144-48-9
EINECS: 205-630-1
UN: 2811
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b08f880f11a25f5a0cf24c7c34a13be729c20290&vid=b08f880f11a25f5a0cf24c7c34a13be729c20290&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR