Hydrazine carbonate ∼7.3 mol/L in water ≥70%
Supplier: TCI America
H0854-25G
H0854-500G
TCH0854-025GEA
17
USD
TCH0854-025G
TCH0854-500G
Hydrazine carbonate ∼7.3 mol/L in water ≥70%
Hydrazine carbonate
CAS Number: 112077-84-6
MDL Number: MFCD00185569
Molecular Formula: CH8N4O2
Molecular Weight: 108.10
Form: Clear Liquid
Color: Colorless
Specific Gravity (20/20): 1.32
Formula:
CH₈N₄O₂ Density: 1.32 (20 °C) |
MDL Number:
MFCD00185569 CAS Number: 112077-84-6 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...