Cetrimonium bromide ≥98.0% for synthesis

Supplier: TCI America
Danger

Synonyms: N-Cetyl-N,N,N-trimethylammonium bromide, Cetyltrimethylammonium bromide, Hexadecyltrimethylammonium bromide, CTAB, (1-Hexadecyl)trimethylammonium bromide, N,N,N-Trimethylhexadecan-1-aminium bromide

H0081-25G H0081-500G H0081-100G
TCH0081-025GEA 24.33 USD
TCH0081-025G TCH0081-500G TCH0081-100G
Cetrimonium bromide ≥98.0% for synthesis
Cetrimonium bromide

CAS Number: 57-09-0 MDL Number: MFCD00011772 Molecular Formula: C19H42BrN Molecular Weight: 364.46 Purity/Analysis Method: 98.0% (T) Form: Crystal Melting point (°C): 243 Flash Point (°C): 244


Product Categories: Materials Chemistry, Cationic Surfactants Other Product Names / Synonyms: Cetyltrimethylammonium Bromide, CTAB

Formula: CH₃(CH₂)₁₅N(Br)(CH₃)₃
Melting Pt: 243 °C
Flash Pt: 244 °C
Storage Temperature: Ambient
MDL Number: MFCD00011772
CAS Number: 57-09-0
EINECS: 200-311-3
UN: 3077
ADR: 9,III
Merck Index: 13,02034
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8793e318ade2880426390ae764ade9cb3e089bd3&vid=8793e318ade2880426390ae764ade9cb3e089bd3&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR