Hecogenin ≥80.0%
Supplier: TCI America
H0003-100MG
H0003-1G
TCH0003-100MGEA
41
USD
TCH0003-100MG
TCH0003-001G
Hecogenin ≥80.0%
Hecogenin
CAS Number: 467-55-0
MDL Number: MFCD00067285
Molecular Formula: C27H42O4
Molecular Weight: 430.63
Purity/Analysis Method:
Formula:
C₂₇H₄₂O₄ MW: 430.63 g/mol |
MDL Number:
MFCD00067285 CAS Number: 467-55-0 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...