D-Glucal ≥97.0%

Supplier: TCI America

Synonyms: 1,5-Anhydro-2-deoxy-D-arabino-hex-1-enitol

G0274-5G G0274-1G
TCG0274-005GEA 198.54 USD
TCG0274-005G TCG0274-1G
D-Glucal ≥97.0%
D-Glucal

CAS Number: 13265-84-4
MDL Number: MFCD00067186
Molecular Formula: C6H10O4
Molecular Weight: 146.14
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Melting point (°C): 62
Specific rotation [a]20/D: -10 deg (C=2, H2O)
Storage Temperature: 0-10°C


Product Categories: Bioscience, Glucose, Glycals
Other Product Names / Synonyms: 1,5-Anhydro-2-deoxy-D-arabino-hex-1-enitol

Formula: C₆H₁₀O₄
MW: 146.14 g/mol
Melting Pt: 62 °C
MDL Number: MFCD00067186
CAS Number: 13265-84-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7bfc1e2ecd853afbb2c22f5cbabcc0f36942765e&vid=7a1e6409207bb083406c1f38e8db00a133806691&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR