Ethyl-2-Amino-4,5,6,7-tetrahydrobenzo[b]thiophene-3-carboxylate ≥98.0% (by titrimetric analysis)
Supplier: TCI America
Synonyms:
2-Amino-3-carbethoxy-4,5-tetramethylenethiophene
, Ethyl 2-amino-4,5,6,7-tetrahydrobenzo[b]thiophene-3-carboxylate
, 2-Amino-3-ethoxycarbonyl-4,5-tetramethylenethiophene
, 2-Amino-4,5,6,7-tetrahydrobenzo[b]thiophene-3-carboxylic Acid Ethyl Ester
CAS Number: 4506-71-2
MDL Number: MFCD00005862
Molecular Formula: C11H15NO2S
Molecular Weight: 225.31
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 115
Other Product Names / Synonyms: 2-Amino-3-carbethoxy-4,5-tetramethylenethiophene, 2-Amino-3-ethoxycarbonyl-4,5-tetramethylenethiophene, 2-Amino-4,5,6,7-tetrahydrobenzo[b]thiophene-3-carboxylic Acid Ethyl Ester
Formula:
C₁₁H₁₅NO₂S MW: 225.31 g/mol Melting Pt: 115 °C |
MDL Number:
MFCD00005862 CAS Number: 4506-71-2 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...