Finasteride ≥98.0%
Supplier: TCI America
Synonyms:
N-tert-Butyl-3-oxo-4-aza-5alpha-androst-1-en-17beta-carboxamide
F0675-1G
F0675-200MG
TCF0675-1GEA
174
USD
TCF0675-1G
TCF0675-200MG
Finasteride ≥98.0%
Finasteride
CAS Number: 98319-26-7
MDL Number: MFCD00869737
Molecular Formula: C23H36N2O2
Molecular Weight: 372.55
Purity/Analysis Method:
Product Categories: Bioscience, Inhibitors (for Pharmacological Research), Antitumor Ingredients, Other Pharmaceutical Ingredients, Other Steroids, Steroids
Other Product Names / Synonyms: N-tert-Butyl-3-oxo-4-aza-5alpha-androst-1-en-17beta-carboxamide
Formula:
C₂₃H₃₆N₂O₂ MW: 372.55 g/mol Melting Pt: 253 °C |
MDL Number:
MFCD00869737 CAS Number: 98319-26-7 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...