1-(5-Fluoro-2,4-dinitrophenyl)-4-methylpiperazine ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: PPZ

F0650-100MG F0650-1G
TCF0650-100MGEA 147 USD
TCF0650-100MG TCF0650-1G
1-(5-Fluoro-2,4-dinitrophenyl)-4-methylpiperazine ≥98.0% (by HPLC, titration analysis)
1-(5-Fluoro-2,4-dinitrophenyl)-4-methylpiperazine

CAS Number: 928830-73-3 Molecular Formula: C11H13FN4O4 Molecular Weight: 284.25 Purity/Analysis Method: 98.0% (HPLC,T) Form: Crystal Melting point (°C): 115 Storage Temperature: 0-10°C


Product Categories: Analytical Science, Derivatization Reagents for MS Other Product Names / Synonyms: PPZ

Formula: C₁₁H₁₃FN₄O₄
MW: 284.25 g/mol
Melting Pt: 115 °C
CAS Number: 928830-73-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ffc78135a53b81973032e25f19a818da67acac74&vid=ffc78135a53b81973032e25f19a818da67acac74&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR