Ethisterone ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
Synonyms:
17alpha-Ethynyltestosterone
E0040-5G
E0040-1G
TCE0040-005GEA
64
USD
TCE0040-005G
TCE0040-001G
Ethisterone ≥98.0% (by HPLC, titration analysis)
Ethisterone
CAS Number: 434-03-7
MDL Number: MFCD00003656
Molecular Formula: C21H28O2
Molecular Weight: 312.45
Purity/Analysis Method:
Product Categories: Synthetic Organic Chemistry, Terminal Alkynes (Click Chemistry), Bioscience, Other Pharmaceutical Ingredients, Steroids, Steroids, Androstanes
Other Product Names / Synonyms: 17alpha-Ethynyltestosterone
Formula:
C₂₁H₂₈O₂ MW: 312.45 g/mol Melting Pt: 272 °C |
MDL Number:
MFCD00003656 CAS Number: 434-03-7 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...