N,N'-Dicyclohexylmorpholine-4-carboximidamide ≥98.0% (by GC, titration analysis)

Supplier: TCI America

D3792-25G D3792-5G
TCD3792-25GEA 97 USD
TCD3792-25G TCD3792-5G
N,N'-Dicyclohexylmorpholine-4-carboximidamide ≥98.0% (by GC, titration analysis)
N,N'-Dicyclohexylmorpholine-4-carboximidamide

CAS Number: 4975-73-9 MDL Number: MFCD00063252 Molecular Formula: C17H31N3O Molecular Weight: 293.46 Purity/Analysis Method: 98.0% (GC,T) Form: Crystal Melting point (°C): 106


Product Categories: Synthetic Organic Chemistry, Nucleic Acid Synthesis Other Product Names / Synonyms: 4-(N,N'-Dicyclohexylamidino)morpholine, N,N'-Dicyclohexylmorpholine-4-carboximidamide, 4-(N,N'-Dicyclohexylcarbamimidoyl)morpholine

Formula: C₁₇H₃₁N₃O
MW: 293.45 g/mol
Melting Pt: 106 °C
MDL Number: MFCD00063252
CAS Number: 4975-73-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4054fd8aa2168862b00a3d91e3e8d4a4f0eda2c8&vid=4054fd8aa2168862b00a3d91e3e8d4a4f0eda2c8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR