7,8-Dihydroxyflavone hydrate ≥98.0%
Supplier: TCI America
Synonyms:
7,8-Dihydroxy-2-phenyl-4H-chromen-4-one, 7,8-Dihydroxyflavone
D1916-1G
D1916-5G
TCD1916-001GEA
100.7
USD
TCD1916-001G
TCD1916-5G
7,8-Dihydroxyflavone hydrate ≥98.0%
7,8-Dihydroxyflavone hydrate
CAS Number: 38183-03-8
MDL Number: MFCD00006836
Molecular Formula: C15H10O4
Molecular Weight: 254.24
Purity/Analysis Method:
Formula:
C₁₅H₁₀O₄ MW: 254.24 g/mol Melting Pt: 245 °C |
MDL Number:
MFCD00006836 CAS Number: 38183-03-8 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...