2-Ethylhexyl-4-(dimethylamino)benzoate ≥98.0%

Supplier: TCI America

Synonyms: 2-Ethylhexyl 4-(dimethylamino)benzoate

D1871-25ML D1871-100ML
TCD1871-025MLEA 38 USD
TCD1871-025ML TCD1871-500ML TCD1871-100ML
2-Ethylhexyl-4-(dimethylamino)benzoate ≥98.0%
2-Ethylhexyl 4-(dimethylamino)benzoate

CAS Number: 21245-02-3 MDL Number: MFCD00017526 Molecular Formula: C17H27NO2 Molecular Weight: 277.41 Purity/Analysis Method: 98.0% (GC,T) Form: Clear Liquid Boiling point (°C): 325 Flash Point (°C): 208 Specific Gravity (20/20): 1.00


Other Product Names / Synonyms: 4-(Dimethylamino)benzoic Acid 2-Ethylhexyl Ester, Octyl 4-(Dimethylamino)benzoate

Formula: C₁₇H₂₇NO₂
MW: 277.40 g/mol
Boiling Pt: 325 °C
Density: 1.00 (20 °C)
Flash Pt: 208 °C
MDL Number: MFCD00017526
CAS Number: 21245-02-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7fdb45057c78a90b005876a0c2ca8d4f465f1388&vid=7fdb45057c78a90b005876a0c2ca8d4f465f1388&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR