Dimethylisopropylchlorosilane ≥90.0%

Supplier: TCI America

Synonyms: Chlorodimethylisopropylsilane, Isopropyldimethylchlorosilane

D1594-25ML D1594-5ML
TCD1594-025MLEA 169 USD
TCD1594-025ML TCD1594-005ML
Dimethylisopropylchlorosilane ≥90.0%
Dimethylisopropylchlorosilane

[Dimethylisopropylsilylating Agent] CAS Number: 3634-56-8 MDL Number: MFCD00009917 Molecular Formula: C5H13ClSi Molecular Weight: 136.69 Purity/Analysis Method: 90.0% (GC) Form: Clear Liquid Boiling point (°C): 112 Flash Point (°C): 15 Specific Gravity (20/20): 0.88


Product Categories: Synthetic Organic Chemistry, Protection & Derivatization Reagents (for Synthesis), Materials Chemistry, Silane Coupling Agents, Analytical Science, Silylation (GC Derivatizing Reagents) Other Product Names / Synonyms: Chlorodimethylisopropylsilane, Isopropyldimethylchlorosilane

Formula: C₅H₁₃ClSi
MW: 136.69 g/mol
Boiling Pt: 112 °C
Density: 0.88 (20 °C)
Flash Pt: 15 °C
MDL Number: MFCD00009917
CAS Number: 3634-56-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cc3ba37f50ad215e753c376201f60b75c1bb85e3&vid=cc3ba37f50ad215e753c376201f60b75c1bb85e3&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 90.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR