Diosgenin ≥95.0%
Supplier: TCI America
Synonyms:
Nitogenin, (2'R,4S,5'R,6aR,6bS,8aS,8bR,9S,11aS,12aS,12bS)-5',6a,8a,9-Tetramethyl-1,3,3',4,4',5,5',6,6a,6b,6',7,8,8a,8b,9,11a,12,12a,12b-icosahydrospiro[naphtho[2',1':4,5]indeno[2,1-b]furan-10,2'-pyran]-4-ol
CAS Number: 512-04-9
MDL Number: MFCD00016887
Molecular Formula: C27H42O3
Molecular Weight: 414.63
Purity/Analysis Method:
Product Categories: Bioscience, Crude Medicine Ingredients for Research and Experimental Use, Steroids (Phytochemicals), Phytosteroids, Steroids
Other Product Names / Synonyms: Nitogenin
Formula:
C₂₇H₄₂O₃ MW: 414.63 g/mol Melting Pt: 205 °C |
MDL Number:
MFCD00016887 CAS Number: 512-04-9 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...