4-Azidobenzoic acid ≥97.0% (by HPLC, titration analysis)

Supplier: TCI America

A0930-25G A0930-5G
TCA0930-025GEA 276.66 USD
TCA0930-025G TCA0930-5G
4-Azidobenzoic acid ≥97.0% (by HPLC, titration analysis)
4-Azidobenzoic acid

CAS Number: 6427-66-3 MDL Number: MFCD00013987 Molecular Formula: C7H5N3O2 Molecular Weight: 163.14 Purity/Analysis Method: 97.0% (HPLC,T) Form: Crystal

Formula: C₇H₅N₃O₂
MW: 163.14 g/mol
MDL Number: MFCD00013987
CAS Number: 6427-66-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=304b87c9648e38ef815d6891c465721bee499538&vid=304b87c9648e38ef815d6891c465721bee499538&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR