Sodium abietate ≥95.0% (by titrimetric analysis)

Supplier: TCI America

A0651-5G A0651-25G
TCA0651-5GEA 98 USD
TCA0651-5G TCA0651-025G
Sodium abietate ≥95.0% (by titrimetric analysis)
Sodium abietate

CAS Number: 14351-66-7 MDL Number: MFCD00058941 Molecular Formula: C20H30O2 Molecular Weight: 324.44 Purity/Analysis Method: 95.0% (T) Form: Crystal


Product Categories: Bioscience, Diterpenes Other Product Names / Synonyms: Abietic Acid Sodium Salt

Formula: C₂₀H₂₉NaO₂
MDL Number: MFCD00058941
CAS Number: 14351-66-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cecded9f5c9e19c7b6815b714e07b6a80ee95222&vid=cecded9f5c9e19c7b6815b714e07b6a80ee95222&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR