Bergapten ≥98.0%

Supplier: TCI America

Synonyms: 5-Methoxypsoralen, 4-Methoxyfuro[3,2-g]benzopyran-7-one, 4-Methoxy-7H-furo[3,2-g]chromen-7-one

B2840-1G B2840-5G
TCB2840-1GEA 191.4 USD
TCB2840-1G TCB2840-5G
Bergapten ≥98.0%
Bergapten

CAS Number: 484-20-8 Molecular Formula: C12H8O4 Molecular Weight: 216.19 Purity/Analysis Method: 98.0% (GC) Form: Crystal Melting point (°C): 191 Lambda max.: 480 nm (MeOH)


Product Categories: Bioscience, Crude Medicine Ingredients for Research and Experimental Use, Phenylpropanoids, Phenylpropanoids (Phytochemicals) Other Product Names / Synonyms: 4-Methoxyfuro[3,2-g]benzopyran-7-one, 5-Methoxypsoralen

Formula: C₁₂H₈O₄
MW: 216.19 g/mol
Melting Pt: 191 °C
MDL Number: MFCD00010272
CAS Number: 484-20-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1c93e99744633361064dc22f4f13712ca8e349ab&vid=1c93e99744633361064dc22f4f13712ca8e349ab&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR