1-Benzyl-3-p-tolyltriazene ≥98.0% (by titrimetric analysis)
Supplier: TCI America
[for Esterification]
CAS Number: 17683-09-9
MDL Number: MFCD00038226
Molecular Formula: C14H15N3
Molecular Weight: 225.30
Purity/Analysis Method:
Product Categories: Synthetic Organic Chemistry, Alkylation (Esterification), Analytical Science, UV Detection (HPLC Labeling Reagents), Esterification & Alkylation (GC Derivatizing Reagents)
Other Product Names / Synonyms: 3-p-Tolyl-1-benzyltriazene
Formula:
C₁₄H₁₅N₃ Melting Pt: 77 °C |
MDL Number:
MFCD00038226 CAS Number: 17683-09-9 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...