Coronene ≥83.0%

Supplier: TCI America

Synonyms: Hexabenzobenzene

C0386-1G C0386-5G
TCC0386-001GEA 192 USD
TCC0386-001G TCC0386-005G
Coronene ≥83.0%
Coronene

CAS Number: 191-07-1
MDL Number: MFCD00004134
Molecular Formula: C24H12
Molecular Weight: 300.36
Purity/Analysis Method: >83.0% (HPLC)
Form: Crystal
Boiling point (°C): 525
Melting point (°C): 440


Product Categories: Materials Chemistry, Molecular Conductors, Nanocarbon Unit Structures (Nanocarbon Materials)
Other Product Names / Synonyms: Hexabenzobenzene

Formula: C₂₄H₁₂
MW: 300.36 g/mol
Boiling Pt: 525
Melting Pt: 440 °C
Density: 1.371 g/cm³ (25 °C)
MDL Number: MFCD00004134
CAS Number: 191-07-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a56f09eb6369d4f43d33169d6ca657637a64c42c&vid=a56f09eb6369d4f43d33169d6ca657637a64c42c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 83.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR