Ferric chloride 2% ethanol

Supplier: Ricca Chemical
Danger

Synonyms: Iron(III) chloride solution

RSD14200-1C
RCSD142-32EA 349 USD
RCSD142-32
Ferric chloride 2% ethanol
Iron(III) chloride solutions
Formula: FeCl₃·6H₂O
MW: 270.3 g/mol
Boiling Pt: ~ 76 °C
Density: 0.8500 g/cm³
Flash Pt: 12.5 °C
Storage Temperature: Ambient
MDL Number: MFCD00149712
CAS Number: 10025-77-1
UN: 1987
ADR: 3 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR