Barium chloride 11,8% (w/v) in aqueous solution

Supplier: Ricca Chemical

R0852000-500A R0852000-1A
RC85216EA 50 USD
RC85216 RC852-32
Barium chloride 11,8% (w/v) in aqueous solution
Barium chloride
Formula: BaCl₂
MW: 208.24 g/mol
Boiling Pt: 1560
Melting Pt: 963
MDL Number: MFCD00003445
CAS Number: 10361-37-2
UN: 1564
ADR: 6.1 III
Merck Index: 12,00998

Order Now

Specification Test Results

Appearance Clear, colorless, odorless
Assay 11.8 ± 0.3 % (w/v)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR