Silver nitrate 0.0192 M (0.0192 N)
Supplier: Ricca Chemical
Synonyms:
Lunar caustic, Silver mononitrate, Nitric acid silver salt
R6910000-1B
R6910000-4B
RC691032EA
103
USD
RC691032
RC6910-1
Silver nitrate 0.0192 M (0.0192 N)
Silver nitrate
1 mL = 1 mg CN-
Formula:
AgNO₃ MW: 169.87 g/mol |
MDL Number:
MFCD00003414 CAS Number: 7761-88-8 UN: 1493 ADR: 5.1,II |
Specification Test Results
Appearance | Clear, colorless, odorless |
Assay at 20 °C (traceable to NIST SRM 999) | 0.01920 ± 0.00001 N at 20.0 °C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...