Potassium hydroxide 0.200 N (N/5)

Supplier: Ricca Chemical

Synonyms: Potash caustic, Potassium hydrate, Caustic potash

R6210000-1A R6210000-20F R6210000-4A
RC6210-32EA 56 USD
RC6210-32 RC6210-5 RC6210-1
Potassium hydroxide 0.200 N (N/5)
Potassium hydroxide
Formula: KOH
MW: 56.11 g/mol
MDL Number: MFCD00003553
CAS Number: 1310-58-3
UN: 1813
ADR: 8,II

Order Now

Specification Test Results

Appearance Clear, colorless, odorless
Assay at 20 °C (traceable to NIST SRM 84) 0.2000 ± 0.0001 N at 20.0 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR