Oxalic acid 6,3% (w/v) in aqueous solution USP Test Solution (TS)
Supplier: Ricca Chemical
|
Danger
|
Synonyms:
Ethanedioic acid
R5466000-1A
R5466000-500A
RC5466-32EA
85
USD
RC5466-32
RC5466-16
Oxalic acid 6,3% (w/v) in aqueous solution USP Test Solution (TS)
Oxalic acid
Formula:
C₂H₂O₄ MW: 90.04 g/mol Boiling Pt: 365 |
MDL Number:
MFCD00002573 CAS Number: 144-62-7 UN: 3265 ADR: 8 III Merck Index: 13,06980 |
Specification Test Results
Appearance | Clear, colorless, odorless |
Refractive Index | 6.3 ± 0.5 % (w/v) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...