Hydrochloric acid R1 250 g/L

Supplier: Ricca Chemical
Danger

R3557000-500A R3557000-1A
RC3557-16EA 50 USD
RC3557-16 RC3557-32
Hydrochloric acid R1 250 g/L
Hydrochloric acid R1
Boiling Pt: ~ 100 °C
Melting Pt: ~ 0 °C
Density: 1.1500 g/cm³
Storage Temperature: Ambient
UN: 1789
ADR: 8,II

Order Now

Specification Test Results

Appearance Clear, colorless, hydrochloric odor
Assay at 20 °C (traceable to NIST SRM 84) 250.0 ± 0.6 g/L HCl


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR