Eriochrome Black T USP Test Solution (TS)
Supplier: Ricca Chemical
|
Danger
|
Synonyms:
1-Naphthalenesulfonic acid, 3-hydroxy-4-[2-(1-hydroxy-2-naphthalenyl)diazenyl]-7-nitro-, sodium salt (1:1)
, 1-Naphthalenesulphonic acid, 3-hydroxy-4-[2-(1-hydroxy-2-naphthalenyl)diazenyl]-7-nitro-, sodium salt (1:1)
, Eriochrome black T, Indicator
, Sodium 3-hydroxy-4-(2-hydroxy-1-naphthylazo)-7-nitro-1-naphthalenesulphonate
, EBT
, Solochrome Black T
, Sodium 3-hydroxy-4-(2-hydroxy-1-naphthylazo)-7-nitro-1-naphthalenesulfonate
R2901000-120C
R2901000-500C
RC2901-16EA
240.42
USD
RC2901-16
RC2901-4
Eriochrome Black T USP Test Solution (TS)
Eriochrome Black T
Formula:
C₂₀H₁₂N₃NaO₇S MW: 461.39 g/mol Boiling Pt: ~ 64.5 °C Density: 0.8000 g/cm³ Flash Pt: ~ 11 °C Storage Temperature: Ambient |
MDL Number:
MFCD00003935 CAS Number: 1787-61-7 UN: 1993 ADR: 3,II |
Specification Test Results
Appearance | Clear, deep red liquid |
Suitability for Use | Wine Red (Excess Ca/Mg) - Blue (Excess EDTA) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...