Bromate-Bromide solution 0.100 N (N/10)

Supplier: Ricca Chemical
Danger

R1170000-4C R1170000-1C R1170000-20F
RC11701EA 269 USD
RC11701 RC117032 RC1170-5
Bromate-Bromide solution 0.100 N (N/10)
Bromate-Bromide

Order Now

Specification Test Results

Appearance Clear, colorless, odorless
Assay at 20 °C (traceable to NIST SRM 136) 0.1000 ± 0.0002 N at 20.0 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR