Flexal™ Nitrile Examination Gloves, Cardinal
Supplier: Cardinal Health
88TN01XS 88TN02S 88TN05XL 88TN06XXL 88TN03M 88TN04L
89194-774CS 825.18 USD
89194-774 89194-776 89194-782 89194-784 89194-778 89194-780
Flexal™ Nitrile Examination Gloves, Cardinal
Gloves
Gloves provide enahnced barrier protection and tactile sensitivity.
  • An alternative to latex
  • Textured fingertips improve grip even when wet
  • Larger box counts
  • Powder-free
  • Thickness: 3.9 mil (finger)