CiDecon® Plus Wipes, Surface Disinfectant Wipes, Decon Labs
Supplier: Avantor
89234-842CS 646.39 USD
89234-842
CiDecon® Plus Wipes, Surface Disinfectant Wipes, Decon Labs
Wipes Cleanroom Wipes Presaturated Cleanroom Wipes
CiDecon® Plus Wipes are pre-moistened hard surface disinfectant wipes for use on hard surfaces.
They come in a convenient 'pop-up' canister, are economical, durable and display superior efficacy against a broad range of pathogenic organisms. Achieves hospital disinfection in 10 minutes and maintains continual residual bacteriostatic activity for up to 6 months (see label for details). FDA compliant for use on medical equipment and instruments. CiDecon is also effective against a wide range of organisms including TB. EPA registered.
Certifications: Meets OSHA's Blood Borne Pathogens Standard.