Chromatography Column with Fritted Disc, Ace Glass Incorporated
Supplier: Ace Glass
5889-20 5889-25 5889-05 5889-10 5889-15 5889-30 5889-35 5889-40
89053-500EA 233.23 USD
89053-500 89053-504 89053-512 89053-514 89053-516 89053-540 89053-582 89053-584
Chromatography Column with Fritted Disc, Ace Glass Incorporated
Chromatography Columns
This chromatography column has a 2 mm bore, 1:5 taper solid PTFE stopcock plug at bottom and a Porosity B (70 to 100 µm) fritted disc sealed in.
The capacity refers only approximate values. Fabricated from borosilicate glass.