(±)-3-Hydroxytetrahydrofuran ≥95%
Supplier: Matrix Scientific
068723-1G
068723-25G
101937-390EA
640
USD
101937-390
101937-394
(±)-3-Hydroxytetrahydrofuran ≥95%
(±)-3-Hydroxytetrahydrofuran
Hazard: Irritant
Formula:
C₄H₈O₂ Boiling Pt: 181 °C Melting Pt: 81 °C Density: 1.09 |
MDL Number:
MFCD00005374 CAS Number: 453-20-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...