1-(6-Chloro-3-nitropyridin-2-yl)azepane ≥95%

Supplier: Matrix Scientific

058215-500MG 058215-1G
101914-252EA 278.46 USD
101914-252 101914-254
1-(6-Chloro-3-nitropyridin-2-yl)azepane ≥95%
1-(6-Chloro-3-nitropyridin-2-yl)azepane

Hazard: Irritant

Formula: C₁₁H₁₄ClN₃O₂
MDL Number: MFCD11206177
CAS Number: 1094400-01-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR