4'-Bromo-2,2,2-trifluoroacetophenone ≥96%
Supplier: Matrix Scientific
055426-5G
055426-25G
101909-238EA
69.23
USD
101909-238
101909-240
4'-Bromo-2,2,2-trifluoroacetophenone ≥96%
4'-Bromo-2,2,2-trifluoroacetophenone
Hazard: Irritant
Formula:
C₈H₄BrF₃O Melting Pt: 26…30 °C |
MDL Number:
MFCD00191862 CAS Number: 16184-89-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...