Ropivacaine hydrochloride
Supplier: Matrix Scientific
Synonyms:
2-Piperidinecarboxamide hydrochloride, Ropivacaine HCl
052755-500MG
052755-2.5G
101900-908EA
140
USD
101900-908
101900-910
Ropivacaine hydrochloride
Ropivacaine hydrochloride
Hazard: Irritant
Formula:
C₁₇H₂₆N₂O·HCl MW: 164.64 g/mol |
MDL Number:
MFCD06671728 CAS Number: 98717-15-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...