Methyl perfluorohexadecanoate ≥95%

Supplier: Matrix Scientific

005783-5G 005783-25G 005783-100G
101789-510EA 53.85 USD
101789-510 101789-512 101789-514
Methyl perfluorohexadecanoate ≥95%
Methyl perfluorohexadecanoate

Hazard: Flammable

Formula: C₁₇H₃F₃₁O₂
Boiling Pt: 167 °C (50 mmHg)
MDL Number: MFCD00236617
CAS Number: 165457-57-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR