8-Iodoquinoline ≥98%
Supplier: Matrix Scientific
069875-1G
069875-25G
101942-336EA
461.54
USD
101942-336
101942-340
8-Iodoquinoline ≥98%
8-Iodoquinoline
Hazard: Irritant
Formula:
C₉H₆IN |
MDL Number:
MFCD08062396 CAS Number: 1006-47-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...