7-Amino-2-methylindazole ≥98%

Supplier: Matrix Scientific

069657-1G 069657-5G
101941-298EA 640 USD
101941-298 101941-300
7-Amino-2-methylindazole ≥98%
7-Amino-2-methylindazole

Hazard: Irritant

Formula: C₈H₉N₃
MDL Number: MFCD03787880
CAS Number: 90223-02-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR