2-Chloroquinoline-3-boronic acid ≥97%

Supplier: Matrix Scientific

069622-1G 069622-5G
101941-128EA 395.38 USD
101941-128 101941-130
2-Chloroquinoline-3-boronic acid ≥97%
2-Chloroquinoline-3-boronic acid

Hazard: Irritant

Formula: C₉H₇BClNO₂
MDL Number: MFCD01114675
CAS Number: 128676-84-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR