2-Amino-4-(trifluoromethyl)oxazole ≥98%

Supplier: Matrix Scientific

Synonyms: 4-(Trifluoromethyl)oxazol-2-amine

069421-500MG 069421-1G 069421-5G
101940-028EA 233.85 USD
101940-028 101940-030 101940-032
2-Amino-4-(trifluoromethyl)oxazole ≥98%
2-Amino-4-(trifluoromethyl)oxazole

Hazard: Irritant

Formula: C₄H₃F₃N₂O
MDL Number: MFCD07777221
CAS Number: 35629-71-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR