Fmoc-2-cyano-L-phenylalanine
Supplier: Matrix Scientific
041926-1G
041926-5G
101853-758EA
538.46
USD
101853-758
101853-760
Fmoc-2-cyano-L-phenylalanine
Fmoc-2-cyano-L-phenylalanine
Hazard: Irritant
Formula:
C₂₅H₂₀N₂O₄ MW: 412.45 g/mol |
MDL Number:
MFCD01863771 CAS Number: 401933-16-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...