N-Boc-2-trifluoromethyl-L-phenylalanine

Supplier: Matrix Scientific

Synonyms: 2-(Trifluoromethyl)-L-phenylalanine, N-BOC protected

041827-1G 041827-5G
101853-362EA 1095.38 USD
101853-362 101853-364
N-Boc-2-trifluoromethyl-L-phenylalanine
N-Boc-2-trifluoromethyl-L-phenylalanine

Hazard: Irritant

Formula: C₁₅H₁₈F₃NO₄
MDL Number: MFCD01862946
CAS Number: 167993-21-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR