Boc-α-methyl-D-aspartic acid

Supplier: Matrix Scientific

041630-100MG 041630-500MG
101852-550EA 536.92 USD
101852-550 101852-552
Boc-α-methyl-D-aspartic acid
Boc-alpha-methyl-D-aspartic acid

Hazard: Irritant

Formula: C₁₀H₁₇NO₆
MDL Number: MFCD02682503
CAS Number: 1217826-44-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR