Nɑ-(tert-Butoxycarbonyl)-Ndelta-carbobenzoxy-L-ornithine

Supplier: Matrix Scientific

Synonyms: Boc-Orn(Z)-OH, Nɑ-Boc-Nδ-Cbz-L-ornithine, Nδ-Benzyloxycarbonyl-Nα-Boc-L-ornithine

041602-1G 041602-5G
101852-434EA 43.08 USD
101852-434 101852-436
Nɑ-(tert-Butoxycarbonyl)-Ndelta-carbobenzoxy-L-ornithine
Nalpha-(tert-Butoxycarbonyl)-Ndelta-carbobenzoxy-L-ornithine

CAS: 2480-93-5 MDL Number: MFCD00038259 Molecular Formula: C18H26N2O6 Molecular Weight: 366,42 Hazard: Irritant

Formula: C₁₈H₂₆N₂O₆
MDL Number: MFCD00038259
CAS Number: 2480-93-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR