Methyl-2-amino-4,5,6,7-tetrahydro-1-benzothiophene-3-carboxylate

Supplier: Matrix Scientific

023866-500MG
101816-932EA 15.38 USD
101816-932
Methyl-2-amino-4,5,6,7-tetrahydro-1-benzothiophene-3-carboxylate
Methyl-2-amino-4,5,6,7-tetrahydro-1-benzothiophene-3-carboxylate

Hazard: Irritant

Formula: C₁₀H₁₃NO₂S
MDL Number: MFCD00130099
CAS Number: 108354-78-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR