4-Amino-α,α,α-trifluoro-o-toluic acid ≥95%

Supplier: Matrix Scientific

Synonyms: 4-Amino-2-(trifluoromethyl)benzoic acid

011208-25G 011208-100G
101800-128EA 106.15 USD
101800-128 102832-832
4-Amino-α,α,α-trifluoro-o-toluic acid ≥95%
4-Amino-α,α,α-trifluoro-o-toluic acid

Hazard: Irritant

Formula: C₈H₆F₃NO₂
MW: 205.14 g/mol
Melting Pt: 185…187 °C
MDL Number: MFCD03407959
CAS Number: 393-06-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR