4-Acetoxythiophene-3-carboxylic acid methyl ester ≥95%

Supplier: Matrix Scientific

058698-1G 058698-5G
101915-464EA 464.62 USD
101915-464 101915-466
4-Acetoxythiophene-3-carboxylic acid methyl ester ≥95%
4-Acetoxythiophene-3-carboxylic acid methyl ester

Purity: 95+% CAS: 65369-31-5 MDL Number: MFCD11227239 Molecular Formula: C8H8O4S Molecular Weight: 200,21 Hazard: Irritant

Formula: C₈H₈O₄S
MDL Number: MFCD11227239
CAS Number: 65369-31-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR