2-Chloro-N-[3-(dimethylamino)propyl]nicotinamide hydrochloride
Supplier: Matrix Scientific
053259-500MG
053259-2.5G
101902-922EA
364.62
USD
101902-922
101902-924
2-Chloro-N-[3-(dimethylamino)propyl]nicotinamide hydrochloride
2-Chloro-N-[3-(dimethylamino)propyl]nicotinamide hydrochloride
Hazard: Irritant
Formula:
C₁₁H₁₇Cl₂N₃O |
MDL Number:
MFCD13562667 CAS Number: 1220037-91-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...