2-[(6-Chloro-2-pyrazinyl)amino]-1-ethanol

Supplier: Matrix Scientific

052866-500MG 052866-2.5G
101901-352EA 364.62 USD
101901-352 101901-354
2-[(6-Chloro-2-pyrazinyl)amino]-1-ethanol
2-[(6-Chloro-2-pyrazinyl)amino]-1-ethanol

Hazard: Irritant

Formula: C₆H₈ClN₃O
MDL Number: MFCD13562417
CAS Number: 1147998-45-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR